6-Fluoro-3-iodo-2-methylpyridine 97%

Supplier: Ambeed
Warning

A388863-1G A388863-5G
76788-826EA 23 USD
76788-826 76788-828
6-Fluoro-3-iodo-2-methylpyridine 97%
6-Fluoro-3-iodo-2-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅FIN
MDL Number: MFCD08277264
CAS Number: 884495-23-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR