Z-Arg(NO2)-OH 98+%

Supplier: Ambeed
Danger

A388098-100G A388098-1G A388098-25G A388098-5G
76788-818EA 166 USD
76788-818 76788-820 76788-822 76788-824
Z-Arg(NO2)-OH 98+%
Z-Arg(NO2)-OH

Precaución: for Research Use Only

Formula: C₁₄H₁₉N₅O₆
MDL Number: MFCD00038092
CAS Number: 2304-98-5
UN: 1325
ADR: 4.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR