2-(2-Ethoxyethoxy)ethyl bromide 98% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilised with potassium carbonate

A387447-100G A387447-1G A387447-25G A387447-5G
76788-798EA 455 USD
76788-798 76788-800 76788-802 76788-804
2-(2-Ethoxyethoxy)ethyl bromide 98% stabilized
2-(2-Ethoxyethoxy)ethyl bromide

Precaución: for Research Use Only

Formula: CH₃CH₂OCH₂CH₂OCH₂CH₂Br
MDL Number: MFCD00051920
CAS Number: 54550-36-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR