Suberic acid 97%

Supplier: Ambeed
Warning

Synonyms: 1,8-Octanedioic acid, Octanedioic acid, 1,6-Hexanedicarboxylic acid

A384765-100G A384765-10G A384765-25G A384765-500G A384765-1000G
76788-788EA 15.92 USD
76788-788 76788-790 76788-792 76788-794 77601-696
Suberic acid 97%
Suberic acid

Precaución: for Research Use Only

Formula: C₈H₁₄O₄
MW: 174.2 g/mol
MDL Number: MFCD00004428
CAS Number: 505-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR