5-Bromo-3-nitropyridine-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A381855-100MG A381855-1G A381855-250MG A381855-5G
76788-776EA 15 USD
76788-776 76788-778 76788-780 77872-598
5-Bromo-3-nitropyridine-2-carboxylic acid 97%
5-Bromo-3-nitropyridine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₃BrN₂O₄
MDL Number: MFCD08061307
CAS Number: 954240-89-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR