Stearyl acrylate 98% stabilised

Supplier: Ambeed
Warning

Synonyms: Octadecyl Acrylate, Acrylic Acid Stearyl Ester, Acrylic Acid Octadecyl Ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A380520-100G A380520-25G A380520-500G
76788-768EA 7 USD
76788-768 76788-770 76788-772
Stearyl acrylate 98% stabilised
Stearyl acrylate

Precaución: for Research Use Only

Formula: C₂₁H₄₀O₂
MW: 324.55 g/mol
MDL Number: MFCD00015091
CAS Number: 4813-57-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR