6-Bromo-3-methylquinazolin-4(3H)-one 98%

Supplier: Ambeed
Warning

A379774-1G A379774-25G A379774-5G
76788-766EA 17 USD
76788-766 77601-670 77668-760
6-Bromo-3-methylquinazolin-4(3H)-one 98%
6-Bromo-3-methylquinazolin-4(3H)-one

Precaución: for Research Use Only

Formula: C₉H₇BrN₂O
MDL Number: MFCD00195328
CAS Number: 57573-59-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR