(6-Bromo-3H-imidazo[4,5-b]pyridin-2-yl)methanol 95%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-2-(hydroxymethyl)-3H-imidazo[4,5-b]pyridine

A376450-100MG A376450-250MG A376450-1G A376450-5G
76788-744EA 38 USD
76788-744 76788-746 77668-744 77872-584
(6-Bromo-3H-imidazo[4,5-b]pyridin-2-yl)methanol 95%
(6-Bromo-3H-imidazo[4,5-b]pyridin-2-yl)methanol

Precaución: for Research Use Only

Formula: C₇H₆BrN₃O
MDL Number: MFCD09750272
CAS Number: 172648-19-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR