6-Bromo-m-toluonitrile 98%

Supplier: Ambeed
Warning

A376063-100G A376063-10G A376063-1G A376063-250MG A376063-25G A376063-5G
76788-728EA 215 USD
76788-728 76788-730 76788-732 76788-734 76788-736 76788-738
6-Bromo-m-toluonitrile 98%
6-Bromo-m-toluonitrile

Precaución: for Research Use Only

Formula: C₈H₆BrN
MDL Number: MFCD11040281
CAS Number: 42872-83-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR