Strontium oxalate 95%

Supplier: Ambeed
Warning

A374698-1000G A374698-500G A374698-100G
76788-724EA 20.16 USD
76788-724 76788-726 77668-740
Strontium oxalate 95%
Strontium oxalate

Precaución: for Research Use Only

Formula: C₂O₄Sr
MW: 175.64 g/mol
MDL Number: MFCD00050803
CAS Number: 814-95-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR