2,6-Diazaspiro[3.4]octane-6-carboxylic acid tert-butyl ester 95%

Supplier: Ambeed
Warning

A373815-100MG A373815-1G A373815-250MG A373815-5G
76788-718EA 34 USD
76788-718 76788-720 76788-722 77872-575
2,6-Diazaspiro[3.4]octane-6-carboxylic acid tert-butyl ester 95%
2,6-Diazaspiro[3.4]octane-6-carboxylic acid tert-butyl ester

Precaución: for Research Use Only

Formula: C₁₁H₂₀N₂O₂
MDL Number: MFCD08234738
CAS Number: 885270-86-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR