1-Thio-β-D-glucose tetraacetate 95%

Supplier: Ambeed
Warning

Synonyms: (2R,3R,4S,5R,6S)-2-(Acetoxymethyl)-6-mercaptotetrahydro-2H-pyran-3,4,5-triyl triacetate

A371264-10G A371264-1G A371264-250MG A371264-25G A371264-5G
76788-670EA 20.16 USD
76788-670 76788-672 76788-674 76788-676 76788-678 76788-680
1-Thio-β-D-glucose tetraacetate 95%
1-Thio-beta-D-glucose tetraacetate

Precaución: for Research Use Only

Formula: C₁₄H₂₀O₉S
MDL Number: MFCD00063271
CAS Number: 19879-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR