4'-(Bromomethyl)-[1,1'-biphenyl]-2-carboxamide 95%

Supplier: Ambeed
Warning

A370363-100MG A370363-1G A370363-250MG
76788-664EA 110.34 USD
76788-664 76788-666 76788-668
4'-(Bromomethyl)-[1,1'-biphenyl]-2-carboxamide 95%
4'-(Bromomethyl)-[1,1'-biphenyl]-2-carboxamide

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrNO
MW: 290.15 g/mol
MDL Number: MFCD11865373
CAS Number: 147404-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR