2-Fluoro-5-(trifluoromethyl)benzophenone 97%

Supplier: Ambeed
Warning

Synonyms: (2-Fluoro-5-(trifluoromethyl)phenyl)(phenyl)methanone

A368919-1G A368919-25G A368919-5G
76788-658EA 17.01 USD
76788-658 76788-660 76788-662
2-Fluoro-5-(trifluoromethyl)benzophenone 97%
2-Fluoro-5-(trifluoromethyl)benzophenone

Precaución: for Research Use Only

Formula: C₁₄H₈F₄O
MDL Number: MFCD00061254
CAS Number: 199292-40-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR