1-Bromo-3-chloro-5-nitrobenzene 97%

Supplier: Ambeed
Warning

A367939-100G A367939-100MG A367939-10G A367939-1G A367939-250MG A367939-25G A367939-5G
76788-636EA 83 USD
76788-636 76788-638 76788-640 76788-642 76788-644 76788-646 76788-648
1-Bromo-3-chloro-5-nitrobenzene 97%
1-Bromo-3-chloro-5-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrClNO₂
MDL Number: MFCD08275708
CAS Number: 219817-43-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR