Tris(2-carboxyethyl)isocyanurate 98%

Supplier: Ambeed
Warning

Synonyms: 3,3',3''-(2,4,6-Trioxo-1,3,5-triazinane-1,3,5-triyl)tripropionic acid, Isocyanuric Acid Tris(2-carboxyethyl) Ester

A367462-25G A367462-5G A367462-100G
76788-630EA 40 USD
76788-630 76788-632 77872-563
Tris(2-carboxyethyl)isocyanurate 98%
Tris(2-carboxyethyl)isocyanurate

Precaución: for Research Use Only

Formula: C₁₂H₁₅N₃O₉
MW: 345.26 g/mol
MDL Number: MFCD00059790
CAS Number: 2904-41-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR