7-Benzyl-5,6,7,8-tetrahydropyrido[3,4-d]pyrimidine-2,4(1H,3H)-dione 95%

Supplier: Ambeed
Warning

A366187-100MG A366187-1G A366187-250MG A366187-5G
76788-614EA 19.1 USD
76788-614 76788-616 76788-618 76788-620
7-Benzyl-5,6,7,8-tetrahydropyrido[3,4-d]pyrimidine-2,4(1H,3H)-dione 95%
7-Benzyl-5,6,7,8-tetrahydropyrido[3,4-d]pyrimidine-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₁₄H₁₅N₃O₂
MW: 257.29 g/mol
MDL Number: MFCD09839775
CAS Number: 62459-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR