4-(Cyanomethyl)-3-fluorobenzonitrile 95%

Supplier: Ambeed
Warning

A365046-1G A365046-250MG
76788-608EA 200.52 USD
76788-608 76788-610
4-(Cyanomethyl)-3-fluorobenzonitrile 95%
4-(Cyanomethyl)-3-fluorobenzonitrile

Precaución: for Research Use Only

Formula: C₉H₅FN₂
MW: 160.14 g/mol
MDL Number: MFCD09813824
CAS Number: 1000516-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR