3,5-Dibromo-N,N-dimethylpyrazinamine 95+%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dibromo-N,N-dimethylpyrazin-2-amine

A362525-1G A362525-250MG A362525-5G
76788-574EA 49 USD
76788-574 76788-576 76788-578
3,5-Dibromo-N,N-dimethylpyrazinamine 95+%
3,5-Dibromo-N,N-dimethylpyrazinamine

Precaución: for Research Use Only

Formula: C₆H₇Br₂N₃
MW: 280.96 g/mol
MDL Number: MFCD09842809
CAS Number: 84539-07-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR