4-Amino-2-chloropyrimidine-5-carboxylic acid 95%

Supplier: Ambeed
Warning

A359978-100MG A359978-1G
76788-552EA 45.62 USD
76788-552 76788-554
4-Amino-2-chloropyrimidine-5-carboxylic acid 95%
4-Amino-2-chloropyrimidine-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₄ClN₃O₂
MW: 173.55 g/mol
MDL Number: MFCD06208432
CAS Number: 773109-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR