4,5-Difluoro-2-methoxyaniline 98%

Supplier: Ambeed
Warning

A359757-1G A359757-250MG
76788-544EA 50 USD
76788-544 77872-545
4,5-Difluoro-2-methoxyaniline 98%
4,5-Difluoro-2-methoxyaniline

Precaución: for Research Use Only

Formula: C₇H₇F₂NO
MDL Number: MFCD09832387
CAS Number: 1017779-71-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR