Tetrahydrofurfuryl methacrylate 98% stabilised
Supplier: Ambeed
|
Warning
|
Synonyms:
(Tetrahydrofuran-2-yl)methyl methacrylate, Methacrylic Acid Tetrahydrofurfuryl Ester
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
A359153-100G
A359153-25G
A359153-500G
76788-530EA
16
USD
76788-530
76788-532
76788-534
Tetrahydrofurfuryl methacrylate 98% stabilised
Tetrahydrofurfuryl methacrylate
Precaución: for Research Use Only
Formula:
C₉H₁₄O₃ MW: 170.21 g/mol |
MDL Number:
MFCD00014515 CAS Number: 2455-24-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...