Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')copper(II) 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Cu(TMHD)2, Copper(II)bis(2,2,6,6-tetramethyl-3,5-heptanedionate), Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)copper(II)
A357063-10G
A357063-1G
A357063-5G
76788-504EA
98.67
USD
76788-504
76788-506
76788-508
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')copper(II) 98%
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')copper(II)
Precaución: for Research Use Only
Formula:
C₂₂H₃₈CuO₄ MW: 430.08 g/mol |
MDL Number:
MFCD00058920 CAS Number: 14040-05-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...