3-Bromo-2-methylanisole 97%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-3-methoxy-2-methylbenzene

A357039-10G A357039-1G A357039-25G A357039-5G A357039-100G
76788-496EA 83.82 USD
76788-496 76788-498 76788-500 76788-502 77668-662
3-Bromo-2-methylanisole 97%
3-Bromo-2-methylanisole

Precaución: for Research Use Only

Formula: C₈H₉BrO
MW: 201.0605 g/mol
MDL Number: MFCD11100991
CAS Number: 31804-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR