Methyl-2-amino-4-methylbenzoate 98%

Supplier: Ambeed
Warning

A356825-1G A356825-250MG A356825-25G A356825-5G
76788-488EA 16 USD
76788-488 76788-490 76788-492 76788-494
Methyl-2-amino-4-methylbenzoate 98%
Methyl-2-amino-4-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂
MDL Number: MFCD09998260
CAS Number: 18595-17-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR