4-(p-Tolyl)-1H-1,2,3-triazole 95%

Supplier: Ambeed
Warning

A356710-100MG A356710-1G
76788-484EA 105 USD
76788-484 76788-486
4-(p-Tolyl)-1H-1,2,3-triazole 95%
4-(p-Tolyl)-1H-1,2,3-triazole

Precaución: for Research Use Only

Formula: C₉H₉N₃
MDL Number: MFCD06251914
CAS Number: 5301-96-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR