(S)-1-(Pyridin-2-yl)ethanamine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(-)-2-[(1S)-1-Aminoethyl]pyridine
A356708-100MG
A356708-10G
A356708-1G
A356708-250MG
A356708-25G
A356708-5G
76788-472EA
28.65
USD
76788-472
76788-474
76788-476
76788-478
76788-480
76788-482
(S)-1-(Pyridin-2-yl)ethanamine 97%
(S)-1-(Pyridin-2-yl)ethanamine
Precaución: for Research Use Only
Formula:
C₇H₁₀N₂ MW: 122.1700 g/mol |
MDL Number:
MFCD08752493 CAS Number: 27854-90-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...