1,4,5,8-Naphthalenetetracarboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: Naphthalene-1,4,5,8-tetracarboxylic acid

A356688-100G A356688-5G A356688-25G
76788-468EA 82.76 USD
76788-468 76788-470 77668-660
1,4,5,8-Naphthalenetetracarboxylic acid 95%
1,4,5,8-Naphthalenetetracarboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₈O₈
MW: 304.21 g/mol
MDL Number: MFCD00004012
CAS Number: 128-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR