N-Succinimidyl 4-Formylbenzoate 97%

Supplier: Ambeed
Warning

Synonyms: Ald-Ph-NHS ester, 2,5-Dioxopyrrolidin-1-yl 4-formylbenzoate, 4-Formylbenzoic Acid N-Succinimidyl Ester

A355420-10MG A355420-1MG A355420-50MG A355420-100MG A355420-1G A355420-250MG A355420-5G
76788-450EA 8 USD
76788-450 76788-452 76788-454 77668-654 77668-656 77872-537 77872-538
N-Succinimidyl 4-Formylbenzoate 97%
N-Succinimidyl 4-Formylbenzoate

Precaución: for Research Use Only

Formula: C₁₂H₉NO₅
MDL Number: MFCD00133733
CAS Number: 60444-78-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR