4-Cyano-5-bromopyrimidine 95%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-4-cyanopyrimidine

A355391-100MG A355391-1G A355391-250MG A355391-5G
76788-442EA 63.66 USD
76788-442 76788-444 76788-446 76788-448
4-Cyano-5-bromopyrimidine 95%
4-Cyano-5-bromopyrimidine

Precaución: for Research Use Only

Formula: C₅H₂BrN₃
MW: 184 g/mol
MDL Number: MFCD09999211
CAS Number: 114969-66-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR