Ethyl-3-aminofuro[2,3-b]pyridine-2-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-aminofuro[2,3-b]pyridine-2-carboxylate

A353916-100MG A353916-250MG A353916-5G
76788-434EA 27.59 USD
76788-434 76788-436 77668-646
Ethyl-3-aminofuro[2,3-b]pyridine-2-carboxylate 95%
Ethyl-3-aminofuro[2,3-b]pyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₃
MW: 206.1980 g/mol
MDL Number: MFCD09837639
CAS Number: 371945-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR