Tetrabutylammonium tetrafluoroborate 98%

Supplier: Ambeed
Danger

Synonyms: Tetrabutylammoniumtetrafluoroborate

A353681-25G A353681-500G
76788-432EA 8.49 USD
76788-432 77551-006
Tetrabutylammonium tetrafluoroborate 98%
Tetrabutylammonium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₁₆H₃₆BF₄N
MW: 329.27 g/mol
MDL Number: MFCD00011634
CAS Number: 429-42-5
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR