1-Methyl-N-carbobenzoxy-L-glutamate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-Carbobenzoxy-L-glutamic Acid 1-Methyl Ester, Z-Glu-OMe, 1-Methyl N-Cbz-L-glutamate, (S)-4-(((Benzyloxy)carbonyl)amino)-5-methoxy-5-oxopentanoic acid, N-Cbz-L-glutamic Acid 1-Methyl Ester
A352399-100G
A352399-10G
A352399-1G
A352399-250MG
A352399-25G
A352399-5G
76788-408EA
98
USD
76788-408
76788-410
76788-412
76788-414
77601-580
77668-622
1-Methyl-N-carbobenzoxy-L-glutamate 98%
1-Methyl-N-carbobenzoxy-L-glutamate
Precaución: for Research Use Only
Formula:
C₁₄H₁₇NO₆ |
MDL Number:
MFCD00083278 CAS Number: 5672-83-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...