1,2-Difluoro-4-(methylsulfonyl)benzene 95%

Supplier: Ambeed
Warning

Synonyms: 3,4-Difluorophenyl methyll sulphone

A351911-1G A351911-5G
76788-404EA 13.8 USD
76788-404 76788-406
1,2-Difluoro-4-(methylsulfonyl)benzene 95%
1,2-Difluoro-4-(methylsulfonyl)benzene

Precaución: for Research Use Only

Formula: C₇H₆F₂O₂S
MW: 192.1800 g/mol
MDL Number: MFCD04037934
CAS Number: 424792-57-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR