5-Chloro-2-methyl-3H-imidazo[4,5-b]pyridine 98%

Supplier: Ambeed
Warning

A350295-100MG A350295-1G A350295-250MG
76788-392EA 35.01 USD
76788-392 76788-394 76788-396
5-Chloro-2-methyl-3H-imidazo[4,5-b]pyridine 98%
5-Chloro-2-methyl-3H-imidazo[4,5-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₆ClN₃
MW: 167.60 g/mol
MDL Number: MFCD08692376
CAS Number: 40851-92-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR