Thiazole-5-carbonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 5-Cyanothiazole

A349862-100MG A349862-1G A349862-250MG
76788-374EA 31 USD
76788-374 76788-376 76788-378
Thiazole-5-carbonitrile 98%
Thiazole-5-carbonitrile

Precaución: for Research Use Only

Formula: C₄H₂N₂S
MDL Number: MFCD06797206
CAS Number: 25742-12-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR