3,5-Difluoro-2-nitroaniline 97%

Supplier: Ambeed
Warning

A349262-1G A349262-250MG A349262-25G A349262-5G
76788-360EA 38.2 USD
76788-360 76788-362 76788-364 76788-366
3,5-Difluoro-2-nitroaniline 97%
3,5-Difluoro-2-nitroaniline

Precaución: for Research Use Only

Formula: C₆H₄F₂N₂O₂
MW: 174.1100 g/mol
MDL Number: MFCD07368747
CAS Number: 361-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR