Tungsten hexacarbonyl 98%

Supplier: Ambeed
Danger

A346359-1G A346359-250MG A346359-5G
76788-332EA 34 USD
76788-332 77601-548 77872-514
Tungsten hexacarbonyl 98%
Tungsten hexacarbonyl

Precaución: for Research Use Only

Formula: C₆O₆W
MDL Number: MFCD00011462
CAS Number: 14040-11-0
UN: 3466
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR