Methyl-5-methyl-4-oxazolecarboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-methyloxazole-4-carboxylate

A346214-1G A346214-250MG
76788-330EA 65 USD
76788-330 77668-590
Methyl-5-methyl-4-oxazolecarboxylate 98%
Methyl 5-methyl-4-oxazolecarboxylate

Precaución: for Research Use Only

Formula: C₆H₇NO₃
MDL Number: MFCD08689502
CAS Number: 41172-57-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR