2'-Hydroxy-3'-methoxyacetophenone 97%
Supplier: Ambeed
|
Warning
|
A345950-100MG
A345950-250MG
76788-328EA
49
USD
76788-328
77601-544
2'-Hydroxy-3'-methoxyacetophenone 97%
2'-Hydroxy-3'-methoxyacetophenone
Precaución: for Research Use Only
Formula:
C₉H₁₀O₃ |
MDL Number:
MFCD12913657 CAS Number: 703-98-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...