N,N-Dimethyl-1-(1H-pyrrolo[2,3-b]pyridin-3-yl)methanamine 98%

Supplier: Ambeed
Danger

Synonyms: 7-Azagramine, 3-(Dimethylaminomethyl)-1H-pyrrolo[2,3-b]pyridine,7-Azagramine,Dimethyl-(1H-pyrrolo[2,3-b]-3-pyridinylmethyl)amine

A345716-100MG A345716-1G A345716-250MG A345716-5G
76788-320EA 21.22 USD
76788-320 76788-322 76788-324 76788-326
N,N-Dimethyl-1-(1H-pyrrolo[2,3-b]pyridin-3-yl)methanamine 98%
N,N-Dimethyl-1-(1H-pyrrolo[2,3-b]pyridin-3-yl)methanamine

Precaución: for Research Use Only

Formula: C₁₀H₁₃N₃
MW: 175.24 g/mol
MDL Number: MFCD01580034
CAS Number: 5654-92-2
UN: 3259
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR