(3-Benzyloxycarbonyl-5-nitrophenyl)boronic acid 96%

Supplier: Ambeed
Warning

Synonyms: 3-Benzyloxycarbonyl-5-nitrophenylboronic acid, (3-((Benzyloxy)carbonyl)-5-nitrophenyl)boronic acid

A342857-1G A342857-250MG A342857-5G
76788-304EA 63 USD
76788-304 76788-306 76788-308
(3-Benzyloxycarbonyl-5-nitrophenyl)boronic acid 96%
(3-Benzyloxycarbonyl-5-nitrophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₂BNO₆
MDL Number: MFCD03411934
CAS Number: 380430-62-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR