cis-3-(Boc-amino)cyclobutanemethanol 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl [cis-3-(hydroxymethyl)cyclobutyl]carbamate, tert-Butyl (cis-3-(hydroxymethyl)cyclobutyl)carbamate, tert-butyl N-[cis-3-(hydroxymethyl)cyclobutyl]carbamate

A342146-250MG A342146-5G
76788-286EA 35.01 USD
76788-286 76788-288
cis-3-(Boc-amino)cyclobutanemethanol 95%
cis-3-(Boc-amino)cyclobutanemethanol

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₃
MW: 201.266 g/mol
MDL Number: MFCD08436074
CAS Number: 142733-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR