3-Pyrroline hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A338353-100G
A338353-1G
A338353-250MG
A338353-5G
A338353-25G
A338353-10G
76788-266EA
603
USD
76788-266
76788-268
76788-270
77571-976
77668-548
77872-495
3-Pyrroline hydrochloride 97%
3-Pyrroline hydrochloride
Precaución: for Research Use Only
Formula:
C₄H₇N·HCl |
MDL Number:
MFCD09991892 CAS Number: 63468-63-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...