tert-Butyl (6-methoxypyridin-3-yl)carbamate 95%

Supplier: Ambeed
Warning

A338127-100MG A338127-1G A338127-250MG A338127-5G
76788-258EA 6 USD
76788-258 76788-260 76788-262 76788-264
tert-Butyl (6-methoxypyridin-3-yl)carbamate 95%
tert-Butyl (6-methoxypyridin-3-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₁₆N₂O₃
MDL Number: MFCD07437873
CAS Number: 183741-80-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR