2,4-Dimethylfluorobenzene 98%

Supplier: Ambeed
Warning

A336025-1G A336025-250MG A336025-5G
76788-240EA 13 USD
76788-240 76788-242 76788-244
2,4-Dimethylfluorobenzene 98%
2,4-Dimethylfluorobenzene

Precaución: for Research Use Only

Formula: C₈H₉F
MDL Number: MFCD00060643
CAS Number: 452-65-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR