(R)-tert-Butyl 2-(2-hydroxyethyl)morpholine-4-carboxylate 97%

Supplier: Ambeed
Warning

A335630-100MG A335630-1G
76788-236EA 147.47 USD
76788-236 76788-238
(R)-tert-Butyl 2-(2-hydroxyethyl)morpholine-4-carboxylate 97%
(R)-tert-Butyl 2-(2-hydroxyethyl)morpholine-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₁NO₄
MW: 231.29 g/mol
MDL Number: MFCD11865208
CAS Number: 136992-21-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR