Strontium bromide hexahydrate 99%

Supplier: Ambeed
Warning

A334594-100G A334594-500G A334594-1000G
76788-222EA 21 USD
76788-222 76788-224 76788-226 77872-485
Strontium bromide hexahydrate 99%
Strontium bromide hexahydrate

Precaución: for Research Use Only

Formula: Br₂Sr·6H₂O
MW: 355.52
MDL Number: MFCD00150361
CAS Number: 7789-53-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR