O,O,O-Tris(4-aminophenyl) phosphorothioate 97%

Supplier: Ambeed
Warning

A334456-1G A334456-5G A334456-25G
76788-218EA 6 USD
76788-218 76788-220 77668-528
O,O,O-Tris(4-aminophenyl) phosphorothioate 97%
O,O,O-Tris(4-aminophenyl) phosphorothioate

Precaución: for Research Use Only

Formula: C₁₈H₁₈N₃O₃PS
MDL Number: MFCD00036100
CAS Number: 52664-35-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR