4-Chloro-6-iodo-7H-pyrrolo[2,3-d]pyrimidine 97%

Supplier: Ambeed
Warning

A334021-100MG A334021-10G A334021-1G A334021-250MG A334021-5G A334021-25G
76788-208EA 6.37 USD
76788-208 76788-210 76788-212 76788-214 76788-216 77668-520
4-Chloro-6-iodo-7H-pyrrolo[2,3-d]pyrimidine 97%
4-Chloro-6-iodo-7H-pyrrolo[2,3-d]pyrimidine

Precaución: for Research Use Only

Formula: C₆H₃ClIN₃
MW: 279.4700 g/mol
MDL Number: MFCD11518919
CAS Number: 876343-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR